Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01410012_m1
          See other IGF2BP3 GE Assays ›
          Gene Symbol:
          Igf2bp3
          Gene Name:
          insulin-like growth factor 2 mRNA binding protein 3
          Gene Aliases:
          Ab2-255, RGD1306512
          Chromosome Location:
          Chr.4: 78744689 - 78879459 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01410012_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Igf2bp3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_003749741.3 XP_003749789.1 11 - 12 1600 69
          XM_006236474.2 XP_006236536.1 11 - 12 1474 69
          XM_006236475.2 XP_006236537.1 9 - 10 1411 69
          XM_008762933.1 XP_008761155.1 11 - 12 1235 69
          GenBank mRNA AY325199.1 - 3 - 4 190 69
          AY724479.1 - 11 - 12 1564 69

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          RNA metabolism protein

          Gene Ontology Categories:

          Process(es)

          mRNA 3'-UTR binding
          poly(A) RNA binding
          mRNA 5'-UTR binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P391156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 877378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243059 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 236291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 786251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 40405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548798 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 92681 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75782 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P391156 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P391156 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P391156 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P391156 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P19259 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1370 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 22371 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33285 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104761 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55804 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32233 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10862431 - -
          Affymetrix Rat Gene 1.0 ST Array 10862429 - -
          Affymetrix Rat Gene 1.0 ST Array 10862439 - -
          Affymetrix Rat Gene 1.0 ST Array 10862433 - -
          Affymetrix Rat Gene 1.0 ST Array 10862438 - -
          Affymetrix Rat Gene 1.0 ST Array 10862428 - -
          Affymetrix Rat Gene 1.0 ST Array 10862441 - -
          Affymetrix Rat Gene 1.0 ST Array 10862432 - -
          Affymetrix Rat Gene 1.0 ST Array 10862443 - -
          Affymetrix Rat Gene 1.0 ST Array 10862435 - -
          Affymetrix Rat Gene 1.0 ST Array 10862440 - -
          Affymetrix Rat Gene 1.0 ST Array 10862427 - -
          Affymetrix Rat Gene 1.0 ST Array 10862436 - -
          Affymetrix Rat Gene 1.0 ST Array 10862425 - -
          Affymetrix Rat Gene 1.0 ST Array 10862442 - -
          Affymetrix Rat Gene 1.0 ST Array 10862426 - -
          Affymetrix Rat Gene 1.0 ST Array 10862437 - -
          Affymetrix Rat Gene 1.0 ST Array 10862434 - -
          Affymetrix Rat Gene 1.0 ST Array 10862430 - -
          Affymetrix Rat Gene 1.1 ST Array 10862424 - -
          Rat Genome 230 2.0 Array 1393202_PM_a_at - -
          Rat Genome 230 2.0 Array 1377824_a_at - -
          Rat Genome 230 2.0 Array 1390860_at - -
          Rat Genome 230 2.0 Array 1390860_PM_at - -
          Rat Genome 230 2.0 Array 1377824_PM_a_at - -
          Rat Genome 230 2.0 Array 1393202_a_at - -
          Affymetrix HT Rat Focus Array 1390860_at - -
          Affymetrix HT Rat Focus Array 1393202_a_at - -
          Affymetrix HT Rat Focus Array 1393202_PM_a_at - -
          Affymetrix HT Rat Focus Array 1377824_PM_a_at - -
          Affymetrix HT Rat Focus Array 1390860_PM_at - -
          Affymetrix HT Rat Focus Array 1377824_a_at - -
          Rat Expression Set 230 1393202_a_at - -
          Rat Expression Set 230 1377824_a_at - -
          Rat Expression Set 230 1377824_PM_a_at - -
          Rat Expression Set 230 1390860_at - -
          Rat Expression Set 230 1393202_PM_a_at - -
          Rat Expression Set 230 1390860_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10862424 - -
          Rat Genome U34 Set rc_AI230709_at - -
          Rat Genome U34 Set rc_AI231489_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21048747 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353191 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0