Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01410038_m1
          See other PPM1K GE Assays ›
          Gene Symbol:
          Ppm1k
          Gene Name:
          protein phosphatase, Mg2+/Mn2+ dependent, 1K
          Gene Aliases:
          RGD1308501
          Chromosome Location:
          Chr.4: 88694370 - 88721374 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01410038_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ppm1k
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107863.1 NP_001101333.1 5 - 6 1144 72
          XM_006236571.2 XP_006236633.1 6 - 7 1475 72
          GenBank mRNA FQ211521.1 - 6 - 7 1267 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400001017.rn.1

          Set Membership:

          Amplicon length between 71 and 85 Protein Phosphatase Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          protein dephosphorylation
          biological_process
          molecular_function
          protein serine/threonine phosphatase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1029435 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205848 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 456079 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653545 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 694916 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 428009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 551275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 214109 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438028 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100935 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 121261 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1029435 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1029435 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1029435 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1029435 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65232 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 82399 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10855856 - -
          Affymetrix Rat Gene 1.0 ST Array 10855854 - -
          Affymetrix Rat Gene 1.0 ST Array 10855859 - -
          Affymetrix Rat Gene 1.0 ST Array 10855858 - -
          Affymetrix Rat Gene 1.0 ST Array 10855855 - -
          Affymetrix Rat Gene 1.0 ST Array 10855857 - -
          Affymetrix Rat Gene 1.1 ST Array 10855853 - -
          Rat Genome 230 2.0 Array 1392921_at - -
          Rat Genome 230 2.0 Array 1394939_at - -
          Rat Genome 230 2.0 Array 1392921_PM_at - -
          Rat Genome 230 2.0 Array 1394939_PM_at - -
          Affymetrix HT Rat Focus Array 1394939_at - -
          Affymetrix HT Rat Focus Array 1394939_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400007238.rn.1 TC0400001017.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400007237.rn.1 TC0400001017.rn.1 RTA TC
          Rat Expression Set 230 1392921_PM_at - -
          Rat Expression Set 230 1394939_at - -
          Rat Expression Set 230 1392921_at - -
          Rat Expression Set 230 1394939_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10855853 - -
          Rat Genome U34 Set rc_AA848311_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21338168 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366955 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline