Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01410138_m1
          See other FKBP9 GE Assays ›
          Gene Symbol:
          Fkbp9
          Gene Name:
          FK506 binding protein 9
          Gene Aliases:
          FKBP-9
          Chromosome Location:
          Chr.4: 87167514 - 87234198 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01410138_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fkbp9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001007646.1 NP_001007647.1 9 - 10 1655 62
          GenBank mRNA BC081961.1 - 9 - 10 1655 7124723 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400000992.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fkbp9

          Gene Ontology Categories:

          Function(s) Process(es)

          protein peptidyl-prolyl isomerization
          protein folding
          chaperone-mediated protein folding
          peptidyl-prolyl cis-trans isomerase activity
          calcium ion binding
          FK506 binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P071515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 680039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 43410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838936 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159183 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569785 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 707933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 641242 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124527 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 162211 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14504 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525967 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 820722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256172 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216511 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52248 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908343 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805622 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 748879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 551675 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101071 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55299 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61559 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108217 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 98268 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 179275 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86811 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P071515 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P071515 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P071515 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P145416 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39842 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41987 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79223 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10137 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10855794 - -
          Affymetrix Rat Gene 1.0 ST Array 10855789 - -
          Affymetrix Rat Gene 1.0 ST Array 10855790 - -
          Affymetrix Rat Gene 1.0 ST Array 10855788 - -
          Affymetrix Rat Gene 1.0 ST Array 10855795 - -
          Affymetrix Rat Gene 1.0 ST Array 10855786 - -
          Affymetrix Rat Gene 1.0 ST Array 10855787 - -
          Affymetrix Rat Gene 1.0 ST Array 10855791 - -
          Affymetrix Rat Gene 1.0 ST Array 10855793 - -
          Affymetrix Rat Gene 1.0 ST Array 10855792 - -
          Affymetrix Rat Gene 1.1 ST Array 10855785 - -
          Rat Genome 230 2.0 Array 1375640_PM_at - -
          Rat Genome 230 2.0 Array 1375640_at - -
          Affymetrix HT Rat Focus Array 1375640_at - -
          Affymetrix HT Rat Focus Array 1375640_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400007127.rn.1 TC0400000992.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400007126.rn.1 TC0400000992.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400007128.rn.1 TC0400000992.rn.1 RTA TC
          Rat Expression Set 230 1375640_PM_at - -
          Rat Expression Set 230 1375640_at - -
          Affymetrix Rat Gene 1.0 ST Array 10855785 - -
          Rat Genome U34 Set rc_AI236786_at - -
          Rat Genome U34 Set rc_AI236301_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22045184 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353562 - -
          RatRef-12 Expression BeadChip ILMN_52852 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline