Search
Search
| Gene Symbol: Nrk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191797.1 | NP_001178726.1 | 3586 | 71 | ||
| XM_008773418.1 | XP_008771640.1 | 3042 | 71 | |||
| XM_008773463.1 | XP_008771685.1 | 1325 | 71 | |||
| XM_008773464.1 | XP_008771686.1 | 5 - 6 | 1324 | 71 | ||
Set Membership: |
|