Search
Search

Serpina7| Gene Symbol: Serpina7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006257307.2 | XP_006257369.1 | 4 - 5 | 1058 | 104 | |
| XM_006257308.2 | XP_006257370.1 | 3 - 4 | 2279 | 104 | ||
| XM_008773465.1 | XP_008771687.1 | 3 - 4 | 2313 | 104 | ||
| XM_008773466.1 | XP_008771688.1 | 4 - 5 | 1051 | 104 | ||
| XM_343823.6 | XP_343824.4 | 3 - 4 | 987 | 104 | ||
| GenBank mRNA | M63991.1 | - | 2 - 3 | 879 | 104 | |
Set Membership: |
|