Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01410602_m1
          See other SHOC2 GE Assays ›
          Gene Symbol:
          Shoc2
          Gene Name:
          soc-2 suppressor of clear homolog (C. elegans)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 274700621 - 274751195 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01410602_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Shoc2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013155.1 NP_001013173.1 1772 70
          GenBank mRNA BC079032.1 - 1772 7114263 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100003833.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Shoc2

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of Ras protein signal transduction
          protein phosphatase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P356962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503173 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583381 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 239012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41403 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 71997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 708259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589100 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 364553 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 685530 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 957166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 451938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554615 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 206891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884600 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 313166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304630 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 578848 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947063 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 676730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 723943 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238889 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123252 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539138 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563968 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 110124 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57915 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13290 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 180448 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108627 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 173246 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8051 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P356962 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P356962 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P356962 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P356962 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89880 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18403 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8736 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85430 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10716155 - -
          Affymetrix Rat Gene 1.0 ST Array 10716156 - -
          Affymetrix Rat Gene 1.1 ST Array 10716154 - -
          Rat Genome 230 2.0 Array 1380435_PM_at - -
          Rat Genome 230 2.0 Array 1380435_at - -
          Affymetrix HT Rat Focus Array 1380435_PM_at - -
          Affymetrix HT Rat Focus Array 1380435_at - -
          Affymetrix Rat Clariom S Assay PSR0100031320.rn.1 TC0100003833.rn.1 RTA TC
          Rat Expression Set 230 1380435_at - -
          Rat Expression Set 230 1380435_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10716154 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22369419 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650845 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline