Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01410778_m1
          See other CYP2C22 GE Assays ›
          Gene Symbol:
          Cyp2c22
          Gene Name:
          cytochrome P450, family 2, subfamily c, polypeptide 22
          Gene Aliases:
          Cyp2c70
          Chromosome Location:
          Chr.1: 259257658 - 259297270 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01410778_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cyp2c22
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_138512.1 NP_612521.1 8 - 9 1315 80
          GenBank mRNA EU861213.1 - 8 - 9 1311 80
          FQ218628.1 - 8 - 9 1312 80
          M58041.1 - 8 - 9 1315 80
          X53477.1 - 8 - 9 1296 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100007672.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cyp2c22

          Panther Classification:

          Molecular Function -

          oxidoreductase oxygenase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          xenobiotic metabolic process
          epoxygenase P450 pathway
          exogenous drug catabolic process
          oxidation-reduction process
          monooxygenase activity
          iron ion binding
          arachidonic acid epoxygenase activity
          steroid hydroxylase activity
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, reduced flavin or flavoprotein as one donor, and incorporation of one atom of oxygen
          oxygen binding
          heme binding
          aromatase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P287842 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89063 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2563 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 44012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352257 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 672938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 713773 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 464516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452072 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 491185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461041 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644134 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 24112 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88441 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93471 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P287842 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P287842 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P287842 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P287842 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700304380 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93364 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86445 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1474 - -
          Agilent Rat cDNA Microarray (G4105A) 700304380 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10730056 - -
          Affymetrix Rat Gene 1.0 ST Array 10730050 - -
          Affymetrix Rat Gene 1.0 ST Array 10730052 - -
          Affymetrix Rat Gene 1.0 ST Array 10730051 - -
          Affymetrix Rat Gene 1.0 ST Array 10730047 - -
          Affymetrix Rat Gene 1.0 ST Array 10730054 - -
          Affymetrix Rat Gene 1.0 ST Array 10730048 - -
          Affymetrix Rat Gene 1.0 ST Array 10730053 - -
          Affymetrix Rat Gene 1.0 ST Array 10730049 - -
          Affymetrix Rat Gene 1.0 ST Array 10730055 - -
          Affymetrix Rat Gene 1.1 ST Array 10730046 - -
          Rat Genome 230 2.0 Array 1387949_PM_at - -
          Rat Genome 230 2.0 Array 1387949_at - -
          Affymetrix HT Rat Focus Array 1387949_at - -
          Affymetrix HT Rat Focus Array 1387949_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100061566.rn.1 TC0100007672.rn.1 RTA TC
          Rat Expression Set 230 1387949_PM_at - -
          Rat Expression Set 230 1387949_at - -
          Affymetrix Rat Gene 1.0 ST Array 10730046 - -
          Rat Genome U34 Set M58041_s_at - -
          Rat Genome U34 Set M58041_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21704364 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373045 - -
          RatRef-12 Expression BeadChip ILMN_56449 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline