Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01410794_m1
          See other OPALIN GE Assays ›
          Gene Symbol:
          Opalin
          Gene Name:
          oligodendrocytic myelin paranodal and inner loop protein
          Gene Aliases:
          Tmem10
          Chromosome Location:
          Chr.1: 260324028 - 260341677 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01410794_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Opalin
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001017386.1 NP_001017386.1 5 - 6 275 83
          XM_006231401.2 XP_006231463.1 3 - 4 1116 83
          GenBank mRNA BC092200.1 - 5 - 6 275 7319643 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100007692.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Opalin

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P133286 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P776423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338890 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 666133 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 777571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 65979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 299839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146326 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 122339 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 144434 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P133286 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P776423 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P776423 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P133286 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P776423 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P133286 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P776423 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P311627 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70687 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14918 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10730154 - -
          Affymetrix Rat Gene 1.0 ST Array 10730151 - -
          Affymetrix Rat Gene 1.0 ST Array 10730150 - -
          Affymetrix Rat Gene 1.0 ST Array 10730149 - -
          Affymetrix Rat Gene 1.0 ST Array 10730153 - -
          Affymetrix Rat Gene 1.0 ST Array 10730152 - -
          Affymetrix Rat Gene 1.1 ST Array 10730148 - -
          Rat Genome 230 2.0 Array 1380329_PM_at - -
          Rat Genome 230 2.0 Array 1380329_at - -
          Affymetrix HT Rat Focus Array 1380329_PM_at - -
          Affymetrix HT Rat Focus Array 1380329_at - -
          Affymetrix Rat Clariom S Assay PSR0100061806.rn.1 TC0100007692.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100061807.rn.1 TC0100007692.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100061808.rn.1 TC0100007692.rn.1 RTA TC
          Rat Expression Set 230 1380329_PM_at - -
          Rat Expression Set 230 1380329_at - -
          Affymetrix Rat Gene 1.0 ST Array 10730148 - -
          Rat Genome U34 Set rc_AI144614_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650340 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline