Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01410806_m1
          See other LZTS2 GE Assays ›
          Gene Symbol:
          Lzts2
          Gene Name:
          leucine zipper, putative tumor suppressor 2
          Gene Aliases:
          Lapser1
          Chromosome Location:
          Chr.1: 264764247 - 264776568 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01410806_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lzts2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014247.2 NP_001014269.2 3 - 4 1366 77
          XM_006231547.2 XP_006231609.1 4 - 5 2416 77
          XM_006231548.2 XP_006231610.1 5 - 6 1617 77
          XM_006231549.2 XP_006231611.1 4 - 5 2845 77
          XM_008760444.1 XP_008758666.1 5 - 6 1608 77
          GenBank mRNA AY928801.1 - 3 - 4 1324 77
          DQ176638.2 - 3 - 4 1324 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100003697.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lzts2

          Gene Ontology Categories:

          Function(s) Process(es)

          kidney development
          mitotic nuclear division
          negative regulation of cell proliferation
          positive regulation of cell death
          Wnt signaling pathway
          negative regulation of Wnt signaling pathway
          microtubule severing
          nuclear export
          spindle midzone assembly
          renal system development
          negative regulation of canonical Wnt signaling pathway
          negative regulation of protein localization to nucleus
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1053550 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P20512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656645 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 948630 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 876253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 214336 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 860963 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71071 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119496 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1053550 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P20512 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P20512 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1053550 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1053550 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P20512 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P466355 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1053550 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41255 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31687 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10715652 - -
          Affymetrix Rat Gene 1.0 ST Array 10715647 - -
          Affymetrix Rat Gene 1.0 ST Array 10715650 - -
          Affymetrix Rat Gene 1.0 ST Array 10715648 - -
          Affymetrix Rat Gene 1.0 ST Array 10715651 - -
          Affymetrix Rat Gene 1.0 ST Array 10715649 - -
          Affymetrix Rat Gene 1.0 ST Array 10715645 - -
          Affymetrix Rat Gene 1.0 ST Array 10715646 - -
          Affymetrix Rat Gene 1.1 ST Array 10715644 - -
          Rat Genome 230 2.0 Array 1389190_at - -
          Rat Genome 230 2.0 Array 1389190_PM_at - -
          Affymetrix HT Rat Focus Array 1389190_PM_at - -
          Affymetrix HT Rat Focus Array 1389190_at - -
          Affymetrix Rat Clariom S Assay PSR0100030091.rn.1 TC0100003697.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100030098.rn.1 TC0100003697.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100030099.rn.1 TC0100003697.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100030096.rn.1 TC0100003697.rn.1 RTA TC
          Rat Expression Set 230 1389190_PM_at - -
          Rat Expression Set 230 1389190_at - -
          Affymetrix Rat Gene 1.0 ST Array 10715644 - -
          Rat Genome U34 Set rc_AI104551_at - -
          Rat Genome U34 Set rc_AI180244_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21294401 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-t4kr2:80/100.66.77.141:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0