Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01410979_m1
          See other POLL GE Assays ›
          Gene Symbol:
          Poll
          Gene Name:
          polymerase (DNA directed), lambda
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 265290337 - 265298840 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01410979_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Poll
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014168.1 NP_001014190.1 7 - 8 1230 56
          XM_006231545.2 XP_006231607.1 6 - 7 1136 56
          GenBank mRNA BC085841.1 - 7 - 8 1230 7135812 56

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100007792.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Poll

          Panther Classification:

          Molecular Function -

          DNA-directed DNA polymerase

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA replication
          nucleotide-excision repair
          biological_process
          DNA biosynthetic process
          molecular_function
          DNA binding
          DNA-directed DNA polymerase activity
          lyase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P143725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238287 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 453991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580702 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126232 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P143725 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P143725 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P143725 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 20185 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10730425 - -
          Affymetrix Rat Gene 1.0 ST Array 10730424 - -
          Affymetrix Rat Gene 1.0 ST Array 10730422 - -
          Affymetrix Rat Gene 1.0 ST Array 10730421 - -
          Affymetrix Rat Gene 1.0 ST Array 10730420 - -
          Affymetrix Rat Gene 1.0 ST Array 10730423 - -
          Affymetrix Rat Gene 1.0 ST Array 10730426 - -
          Affymetrix Rat Gene 1.0 ST Array 10730427 - -
          Affymetrix Rat Gene 1.1 ST Array 10730419 - -
          Rat Genome 230 2.0 Array 1389268_at - -
          Rat Genome 230 2.0 Array 1389268_PM_at - -
          Affymetrix HT Rat Focus Array 1389268_at - -
          Affymetrix HT Rat Focus Array 1389268_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100062790.rn.1 TC0100007792.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100062789.rn.1 TC0100007792.rn.1 RTA TC
          Rat Expression Set 230 1389268_PM_at - -
          Rat Expression Set 230 1389268_at - -
          Affymetrix Rat Gene 1.0 ST Array 10730419 - -
          Rat Genome U34 Set rc_AI045911_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20899687 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364120 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline