Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024882.1 | NP_001020053.1 | 1 - 2 | 36 | 79 | |
XM_006250296.2 | XP_006250358.1 | 1 - 2 | 131 | 79 | ||
GenBank mRNA | BC098058.1 | - | 1 - 2 | 36 | 7457890 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |