Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01412340_m1
          See other IGSF8 GE Assays ›
          Gene Symbol:
          Igsf8
          Gene Name:
          immunoglobulin superfamily, member 8
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 90692507 - 90700795 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01412340_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Igsf8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014787.1 NP_001014787.1 3 - 4 506 153
          XM_006250286.2 XP_006250348.1 3 - 4 665 153
          XM_006250288.2 XP_006250350.1 2 - 3 607 153
          XM_006250289.2 XP_006250351.1 2 - 3 444 153
          GenBank mRNA BC092192.1 - 3 - 4 506 7312924 153

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300000785.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Igsf8

          Panther Classification:

          Molecular Function -

          immunoglobulin receptor superfamily

          Gene Ontology Categories:

          Function(s)

          regulation of cell motility

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P14103 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560369 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655744 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162467 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P14103 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P14103 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P14103 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46719 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10765734 - -
          Affymetrix Rat Gene 1.0 ST Array 10765736 - -
          Affymetrix Rat Gene 1.0 ST Array 10765739 - -
          Affymetrix Rat Gene 1.0 ST Array 10765735 - -
          Affymetrix Rat Gene 1.0 ST Array 10765738 - -
          Affymetrix Rat Gene 1.0 ST Array 10765731 - -
          Affymetrix Rat Gene 1.0 ST Array 10765732 - -
          Affymetrix Rat Gene 1.0 ST Array 10765733 - -
          Affymetrix Rat Gene 1.1 ST Array 10765730 - -
          Rat Genome 230 2.0 Array 1373793_PM_at - -
          Rat Genome 230 2.0 Array 1373793_at - -
          Affymetrix HT Rat Focus Array 1373793_at - -
          Affymetrix HT Rat Focus Array 1373793_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1300005615.rn.1 TC1300000785.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300005620.rn.1 TC1300000785.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300005617.rn.1 TC1300000785.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300005612.rn.1 TC1300000785.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300005618.rn.1 TC1300000785.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300005619.rn.1 TC1300000785.rn.1 RTA TC
          Rat Expression Set 230 1373793_PM_at - -
          Rat Expression Set 230 1373793_at - -
          Affymetrix Rat Gene 1.0 ST Array 10765730 - -
          Rat Genome U34 Set rc_AA858677_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22418478 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline