Search
Search
| Gene Symbol: Birc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008762598.1 | XP_008760820.1 | 1 - 2 | 1228 | 92 | |
| XM_008762599.1 | XP_008760821.1 | 1 - 2 | 1228 | 92 | ||
| XM_008762600.1 | XP_008760822.1 | 1 - 2 | 1228 | 92 | ||
Set Membership: |
|