Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01412727_m1
          See other RNF216 GE Assays ›
          Gene Symbol:
          Rnf216
          Gene Name:
          ring finger protein 216
          Gene Aliases:
          RGD1565135
          Chromosome Location:
          Chr.12: 13508429 - 13631689 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01412727_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rnf216
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107122.1 NP_001100592.1 11 - 12 1952 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200000240.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Apoptosis Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of interferon-beta production
          proteasome-mediated ubiquitin-dependent protein catabolic process
          regulation of defense response to virus by host
          protein K48-linked ubiquitination
          zinc ion binding
          ligase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P617381 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P063903 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 972887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 245209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316689 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119714 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 611722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 728920 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967530 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 703085 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600331 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 937690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 184972 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 51239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530200 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538687 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 846896 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 464718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377019 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 241448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 666589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671235 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 326073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640489 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 528206 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 687618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449952 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 425939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 303028 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176676 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 464579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 196296 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 687130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584086 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634804 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205978 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41947 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 11547 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 65904 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 35039 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 11627 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 104231 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 97428 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 18483 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67939 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 131679 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154576 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5566 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P617381 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P063903 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P617381 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P063903 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P617381 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P063903 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P527286 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P617381 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 59631 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74350 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4081 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10756791 - -
          Affymetrix Rat Gene 1.0 ST Array 10756793 - -
          Affymetrix Rat Gene 1.0 ST Array 10756795 - -
          Affymetrix Rat Gene 1.0 ST Array 10756789 - -
          Affymetrix Rat Gene 1.0 ST Array 10756784 - -
          Affymetrix Rat Gene 1.0 ST Array 10756787 - -
          Affymetrix Rat Gene 1.0 ST Array 10756792 - -
          Affymetrix Rat Gene 1.0 ST Array 10756797 - -
          Affymetrix Rat Gene 1.0 ST Array 10756790 - -
          Affymetrix Rat Gene 1.0 ST Array 10756798 - -
          Affymetrix Rat Gene 1.0 ST Array 10756785 - -
          Affymetrix Rat Gene 1.0 ST Array 10756788 - -
          Affymetrix Rat Gene 1.0 ST Array 10756794 - -
          Affymetrix Rat Gene 1.0 ST Array 10756786 - -
          Affymetrix Rat Gene 1.0 ST Array 10756796 - -
          Affymetrix Rat Gene 1.1 ST Array 10756783 - -
          Rat Genome 230 2.0 Array 1376608_PM_at - -
          Rat Genome 230 2.0 Array 1376608_at - -
          Affymetrix HT Rat Focus Array 1376608_at - -
          Affymetrix HT Rat Focus Array 1376608_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1200001738.rn.1 TC1200000240.rn.1 RTA TC
          Rat Expression Set 230 1376608_PM_at - -
          Rat Expression Set 230 1376608_at - -
          Affymetrix Rat Gene 1.0 ST Array 10756783 - -
          Rat Genome U34 Set rc_AI112166_at - -
          Rat Genome U34 Set rc_AI236535_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22108609 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350644 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline