Search
Search
| Gene Symbol: Pan3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006248842.2 | XP_006248904.1 | 14 - 15 | 2418 | 74 | |
| XM_006248844.2 | XP_006248906.1 | 14 - 15 | 2418 | 74 | ||
| XM_006248845.2 | XP_006248907.1 | 14 - 15 | 2421 | 74 | ||
| XM_346914.7 | XP_346915.4 | 13 - 14 | 2255 | 74 | ||
Set Membership: |
|