Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01412968_m1
          See other ZDHHC4 GE Assays ›
          Gene Symbol:
          Zdhhc4
          Gene Name:
          zinc finger, DHHC-type containing 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 13284527 - 13298519 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01412968_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zdhhc4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013123.1 NP_001013141.1 5 - 6 918 128
          XM_006248876.2 XP_006248938.1 7 - 8 998 128
          XM_008768962.1 XP_008767184.1 8 - 9 1122 128
          GenBank mRNA AY886523.1 - 6 - 7 923 128
          BC089831.1 - 5 - 6 918 7323958 128
          FQ214800.1 - 7 - 8 1004 128

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200000230.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zdhhc4

          Gene Ontology Categories:

          Function(s) Process(es)

          metabolic process
          zinc ion binding
          protein-cysteine S-palmitoyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1045141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516385 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940704 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 288295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466126 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40639 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1045141 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1045141 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1045141 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1045141 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6864 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10756739 - -
          Affymetrix Rat Gene 1.0 ST Array 10756737 - -
          Affymetrix Rat Gene 1.0 ST Array 10756734 - -
          Affymetrix Rat Gene 1.0 ST Array 10756735 - -
          Affymetrix Rat Gene 1.0 ST Array 10756736 - -
          Affymetrix Rat Gene 1.0 ST Array 10756738 - -
          Affymetrix Rat Gene 1.0 ST Array 10756733 - -
          Affymetrix Rat Gene 1.0 ST Array 10756732 - -
          Affymetrix Rat Gene 1.1 ST Array 10756731 - -
          Rat Genome 230 2.0 Array 1374461_PM_at - -
          Rat Genome 230 2.0 Array 1374461_at - -
          Affymetrix HT Rat Focus Array 1374461_at - -
          Affymetrix HT Rat Focus Array 1374461_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1200001633.rn.1 TC1200000230.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200001632.rn.1 TC1200000230.rn.1 RTA TC
          Rat Expression Set 230 1374461_PM_at - -
          Rat Expression Set 230 1374461_at - -
          Affymetrix Rat Gene 1.0 ST Array 10756731 - -
          Rat Genome U34 Set rc_AA957719_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21389829 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline