Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01413246_m1
          See other IGFBP7 GE Assays ›
          Gene Symbol:
          Igfbp7
          Gene Name:
          insulin-like growth factor binding protein 7
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 33010300 - 33070193 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01413246_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Igfbp7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013048.1 NP_001013066.1 2 - 3 595 101
          GenBank mRNA BC086582.1 - 2 - 3 595 7307584 101

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400000368.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Igfbp7

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cell growth
          cell adhesion
          embryo implantation
          response to heat
          response to organic cyclic compound
          response to retinoic acid
          cellular response to hormone stimulus
          inner ear development
          regulation of steroid biosynthetic process
          response to cortisol
          molecular_function
          insulin-like growth factor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P048187 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457063 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 685541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 162225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 330042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 748697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450108 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 61214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 895124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958768 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 367248 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 423652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128572 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883252 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278943 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4110 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 742952 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265951 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 877625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796455 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564776 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 326984 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161906 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42023 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158476 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89598 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158996 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 123067 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7934 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P048187 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P048187 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P048187 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85410 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6871 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88720 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75876 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74789 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10772173 - -
          Affymetrix Rat Gene 1.0 ST Array 10772172 - -
          Affymetrix Rat Gene 1.0 ST Array 10772174 - -
          Affymetrix Rat Gene 1.0 ST Array 10772170 - -
          Affymetrix Rat Gene 1.0 ST Array 10772175 - -
          Affymetrix Rat Gene 1.0 ST Array 10772171 - -
          Affymetrix Rat Gene 1.1 ST Array 10772169 - -
          Rat Genome 230 2.0 Array 1371357_at - -
          Rat Genome 230 2.0 Array 1371357_PM_at - -
          Affymetrix HT Rat Focus Array 1371357_PM_at - -
          Affymetrix HT Rat Focus Array 1371357_at - -
          Affymetrix Rat Clariom S Assay PSR1400002912.rn.1 TC1400000368.rn.1 RTA TC
          Rat Expression Set 230 1371357_at - -
          Rat Expression Set 230 1371357_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10772169 - -
          Rat Genome U34 Set rc_AI233246_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22237389 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368086 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline