Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01413379_g1
          See other LIAS GE Assays ›
          Gene Symbol:
          Lias
          Gene Name:
          lipoic acid synthetase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 44507217 - 44524289 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01413379_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lias
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012037.1 NP_001012037.1 1 - 2 109 70
          XM_008770155.1 XP_008768377.1 1 - 2 145 70
          XM_008770156.1 XP_008768378.1 1 - 2 146 70
          GenBank mRNA BC083708.1 - 1 - 2 109 7190554 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400001778.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lias

          Gene Ontology Categories:

          Function(s) Process(es)

          neural tube closure
          inflammatory response
          response to oxidative stress
          biological_process
          lipoate biosynthetic process
          protein lipoylation
          response to lipopolysaccharide
          molecular_function
          lipoate synthase activity
          metal ion binding
          4 iron, 4 sulfur cluster binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P054411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540067 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955572 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 547977 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 236858 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41437 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107843 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41058 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P054411 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P054411 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P054411 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P187320 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P380992 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27181 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10776813 - -
          Affymetrix Rat Gene 1.0 ST Array 10776817 - -
          Affymetrix Rat Gene 1.0 ST Array 10776821 - -
          Affymetrix Rat Gene 1.0 ST Array 10776814 - -
          Affymetrix Rat Gene 1.0 ST Array 10776816 - -
          Affymetrix Rat Gene 1.0 ST Array 10776815 - -
          Affymetrix Rat Gene 1.0 ST Array 10776812 - -
          Affymetrix Rat Gene 1.0 ST Array 10776818 - -
          Affymetrix Rat Gene 1.0 ST Array 10776819 - -
          Affymetrix Rat Gene 1.0 ST Array 10776820 - -
          Affymetrix Rat Gene 1.1 ST Array 10776811 - -
          Rat Genome 230 2.0 Array 1389154_at - -
          Rat Genome 230 2.0 Array 1389154_PM_at - -
          Affymetrix HT Rat Focus Array 1389154_PM_at - -
          Affymetrix HT Rat Focus Array 1389154_at - -
          Affymetrix Rat Clariom S Assay PSR1400013071.rn.1 TC1400001778.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400013072.rn.1 TC1400001778.rn.1 RTA TC
          Rat Expression Set 230 1389154_at - -
          Rat Expression Set 230 1389154_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10776811 - -
          Rat Genome U34 Set rc_AI237159_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_2040743 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376103 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline