Search
Search
| Gene Symbol: Atp8a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006250974.2 | XP_006251036.1 | 32 - 33 | 3260 | 61 | |
| XM_006250975.2 | XP_006251037.1 | 32 - 33 | 3260 | 61 | ||
| XM_006250976.2 | XP_006251038.1 | 31 - 32 | 3245 | 61 | ||
| XM_006250977.2 | XP_006251039.1 | 31 - 32 | 3215 | 61 | ||
| XM_006250980.2 | XP_006251042.1 | 28 - 29 | 2746 | 61 | ||
| XM_008770164.1 | XP_008768386.1 | 32 - 33 | 3290 | 61 | ||
| XM_008770165.1 | XP_008768387.1 | 29 - 30 | 2998 | 61 | ||
Set Membership: |
|