Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01413619_m1
          See other KLF3 GE Assays ›
          Gene Symbol:
          Klf3
          Gene Name:
          Kruppel-like factor 3 (basic)
          Gene Aliases:
          Bklf, Tef-2
          Chromosome Location:
          Chr.14: 45142769 - 45173753 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01413619_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Klf3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001105742.1 NP_001099212.1 4 - 5 923 116
          XM_006250995.2 XP_006251057.1 4 - 5 6320 116
          XM_008770134.1 XP_008768356.1 4 - 5 837 116

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400001791.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          cellular response to peptide
          nucleic acid binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P461640 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P573441 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783794 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27894 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582267 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 339415 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 333069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 676645 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 16430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780393 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 361456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263282 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 19663 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 168267 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174808 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P573441 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P461640 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P573441 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P461640 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P573441 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P461640 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P461640 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10776909 - -
          Affymetrix Rat Gene 1.0 ST Array 10776908 - -
          Affymetrix Rat Gene 1.0 ST Array 10776905 - -
          Affymetrix Rat Gene 1.0 ST Array 10776907 - -
          Affymetrix Rat Gene 1.0 ST Array 10776906 - -
          Affymetrix Rat Gene 1.1 ST Array 10776904 - -
          Rat Genome 230 2.0 Array 1378332_at - -
          Rat Genome 230 2.0 Array 1393150_at - -
          Rat Genome 230 2.0 Array 1389479_PM_at - -
          Rat Genome 230 2.0 Array 1389479_at - -
          Rat Genome 230 2.0 Array 1378332_PM_at - -
          Rat Genome 230 2.0 Array 1393150_PM_at - -
          Affymetrix HT Rat Focus Array 1393150_at - -
          Affymetrix HT Rat Focus Array 1389479_at - -
          Affymetrix HT Rat Focus Array 1378332_PM_at - -
          Affymetrix HT Rat Focus Array 1389479_PM_at - -
          Affymetrix HT Rat Focus Array 1393150_PM_at - -
          Affymetrix HT Rat Focus Array 1378332_at - -
          Affymetrix Rat Clariom S Assay PSR1400013198.rn.1 TC1400001791.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400013199.rn.1 TC1400001791.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400013200.rn.1 TC1400001791.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400013201.rn.1 TC1400001791.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400013203.rn.1 TC1400001791.rn.1 RTA TC
          Rat Expression Set 230 1389479_at - -
          Rat Expression Set 230 1389479_PM_at - -
          Rat Expression Set 230 1378332_PM_at - -
          Rat Expression Set 230 1393150_PM_at - -
          Rat Expression Set 230 1378332_at - -
          Rat Expression Set 230 1393150_at - -
          Affymetrix Rat Gene 1.0 ST Array 10776904 - -
          Rat Genome U34 Set rc_AA956804_at - -
          Rat Genome U34 Set rc_AI059323_at - -
          Rat Genome U34 Set rc_AI233750_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20819488 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354006 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-t4kr2:80/100.66.77.141:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0