Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01413725_m1
          See other DCLRE1A GE Assays ›
          Gene Symbol:
          Dclre1a
          Gene Name:
          DNA cross-link repair 1A
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 277335792 - 277355691 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01413725_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dclre1a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106201.1 NP_001099671.1 6 - 7 2753 71
          XM_006231634.1 XP_006231696.1 7 - 8 3429 71
          XM_006231635.2 XP_006231697.1 7 - 8 3367 71
          XM_008760523.1 XP_008758745.1 7 - 8 3429 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100007946.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          DNA metabolism protein

          Gene Ontology Categories:

          Function(s)

          nucleotide-excision repair

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P087920 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 249885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631455 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580523 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 414181 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 626406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107907 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124651 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 696779 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205529 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807380 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 36309 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 43302 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P087920 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P087920 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P087920 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1050352 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10731002 - -
          Affymetrix Rat Gene 1.0 ST Array 10731007 - -
          Affymetrix Rat Gene 1.0 ST Array 10731001 - -
          Affymetrix Rat Gene 1.0 ST Array 10731008 - -
          Affymetrix Rat Gene 1.0 ST Array 10731004 - -
          Affymetrix Rat Gene 1.0 ST Array 10731005 - -
          Affymetrix Rat Gene 1.0 ST Array 10731000 - -
          Affymetrix Rat Gene 1.0 ST Array 10731006 - -
          Affymetrix Rat Gene 1.0 ST Array 10731003 - -
          Affymetrix Rat Gene 1.1 ST Array 10730999 - -
          Rat Genome 230 2.0 Array 1396044_at - -
          Rat Genome 230 2.0 Array 1383495_at - -
          Rat Genome 230 2.0 Array 1383495_PM_at - -
          Rat Genome 230 2.0 Array 1396044_PM_at - -
          Affymetrix HT Rat Focus Array 1383495_at - -
          Affymetrix HT Rat Focus Array 1383495_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100064005.rn.1 TC0100007946.rn.1 RTA TC
          Rat Expression Set 230 1383495_at - -
          Rat Expression Set 230 1383495_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10730999 - -
          Rat Genome U34 Set rc_AA944332_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22035959 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_2039608 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372971 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline