Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01413733_m1
          See other WNT8B GE Assays ›
          Gene Symbol:
          Wnt8b
          Gene Name:
          wingless-type MMTV integration site family, member 8B
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 264244200 - 264264330 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01413733_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Wnt8b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106359.1 NP_001099829.1 2 - 3 181 70
          XM_006231442.2 XP_006231504.1 3 - 4 337 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100003683.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          positive regulation of gene expression
          negative regulation of gene expression
          Wnt signaling pathway
          neuron differentiation
          cell fate commitment
          molecular_function
          frizzled binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P098474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57237 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 2179 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P098474 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P098474 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P098474 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P557726 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10715564 - -
          Affymetrix Rat Gene 1.0 ST Array 10715565 - -
          Affymetrix Rat Gene 1.0 ST Array 10715563 - -
          Affymetrix Rat Gene 1.0 ST Array 10715561 - -
          Affymetrix Rat Gene 1.0 ST Array 10715562 - -
          Affymetrix Rat Gene 1.1 ST Array 10715559 - -
          Rat Genome 230 2.0 Array 1386122_at - -
          Rat Genome 230 2.0 Array 1386122_PM_at - -
          Affymetrix HT Rat Focus Array 1386122_PM_at - -
          Affymetrix HT Rat Focus Array 1386122_at - -
          Affymetrix Rat Clariom S Assay PSR0100029925.rn.1 TC0100003683.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100029923.rn.1 TC0100003683.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100029924.rn.1 TC0100003683.rn.1 RTA TC
          Rat Expression Set 230 1386122_PM_at - -
          Rat Expression Set 230 1386122_at - -
          Affymetrix Rat Gene 1.0 ST Array 10715559 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21613991 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355107 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-t4kr2:80/100.66.77.141:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0