Search
Search

Cyp2c23| Gene Symbol: Cyp2c23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031839.3 | NP_114027.2 | 1 - 2 | 207 | 60 | |
| GenBank mRNA | BC078707.1 | - | 1 - 2 | 207 | 7110580 | 60 |
| U04733.1 | - | 1 - 2 | 188 | 60 | ||
Set Membership: |
|