Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01413762_m1
          See other PSD GE Assays ›
          Gene Symbol:
          Psd
          Gene Name:
          pleckstrin and Sec7 domain containing
          Gene Aliases:
          EFA6
          Chromosome Location:
          Chr.1: 266059331 - 266074781 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01413762_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Psd
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_134370.2 NP_599197.2 17 - 18 3130 60
          XM_006231432.2 XP_006231494.1 17 - 18 3046 60
          XM_008760393.1 XP_008758615.1 16 - 17 3042 60
          XM_008760394.1 XP_008758616.1 17 - 18 3235 60
          XM_008760395.1 XP_008758617.1 17 - 18 3237 60
          XM_008760396.1 XP_008758618.1 17 - 18 3026 60
          GenBank mRNA AB040468.1 - 14 - 15 2121 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100007814.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Psd

          Panther Classification:

          Molecular Function -

          guanyl-nucleotide exchange factor

          Gene Ontology Categories:

          Function(s) Process(es)

          guanine nucleotide transport
          guanine transport
          vesicle-mediated transport
          neuron differentiation
          regulation of ARF protein signal transduction
          positive regulation of GTPase activity
          guanine nucleotide transmembrane transporter activity
          ARF guanyl-nucleotide exchange factor activity
          phospholipid binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P026379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607187 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132147 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 100517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 40046 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 112218 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P026379 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P026379 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P026379 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P13247 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87360 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10730546 - -
          Affymetrix Rat Gene 1.0 ST Array 10730554 - -
          Affymetrix Rat Gene 1.0 ST Array 10730541 - -
          Affymetrix Rat Gene 1.0 ST Array 10730557 - -
          Affymetrix Rat Gene 1.0 ST Array 10730544 - -
          Affymetrix Rat Gene 1.0 ST Array 10730545 - -
          Affymetrix Rat Gene 1.0 ST Array 10730540 - -
          Affymetrix Rat Gene 1.0 ST Array 10730549 - -
          Affymetrix Rat Gene 1.0 ST Array 10730547 - -
          Affymetrix Rat Gene 1.0 ST Array 10730555 - -
          Affymetrix Rat Gene 1.0 ST Array 10730543 - -
          Affymetrix Rat Gene 1.0 ST Array 10730558 - -
          Affymetrix Rat Gene 1.0 ST Array 10730552 - -
          Affymetrix Rat Gene 1.0 ST Array 10730553 - -
          Affymetrix Rat Gene 1.0 ST Array 10730542 - -
          Affymetrix Rat Gene 1.0 ST Array 10730551 - -
          Affymetrix Rat Gene 1.0 ST Array 10730550 - -
          Affymetrix Rat Gene 1.0 ST Array 10730556 - -
          Affymetrix Rat Gene 1.1 ST Array 10730539 - -
          Rat Genome 230 2.0 Array 1387572_PM_at - -
          Rat Genome 230 2.0 Array 1387572_at - -
          Affymetrix HT Rat Focus Array 1387572_at - -
          Affymetrix HT Rat Focus Array 1387572_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100063063.rn.1 TC0100007814.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100063064.rn.1 TC0100007814.rn.1 RTA TC
          Rat Expression Set 230 1387572_PM_at - -
          Rat Expression Set 230 1387572_at - -
          Affymetrix Rat Gene 1.0 ST Array 10730539 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21570913 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_2040891 - -
          RatRef-12 Expression BeadChip ILMN_62376 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline