Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01413889_g1
          See other CXCL10 GE Assays ›
          Gene Symbol:
          Cxcl10
          Gene Name:
          chemokine (C-X-C motif) ligand 10
          Gene Aliases:
          IP-10, Scyb10
          Chromosome Location:
          Chr.14: 17210733 - 17212930 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01413889_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cxcl10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_139089.1 NP_620789.1 2 - 3 278 106
          GenBank mRNA BC058444.1 - 2 - 3 293 6918440 106
          FQ225217.1 - 2 - 3 292 106
          FQ225221.1 - 2 - 3 292 106
          FQ225582.1 - 2 - 3 289 106
          FQ225670.1 - 2 - 3 292 106
          FQ226479.1 - 2 - 3 289 106
          FQ231406.1 - 2 - 3 292 106
          FQ232883.1 - 2 - 3 292 106
          FQ233441.1 - 2 - 3 292 106
          FQ233623.1 - 2 - 3 292 106
          FQ233884.1 - 2 - 3 292 106
          U17035.1 - 2 - 3 278 106
          U22520.1 - 2 - 3 277 106

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400000252.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cxcl10

          Panther Classification:

          Molecular Function -

          chemokine

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of leukocyte chemotaxis
          chemotaxis
          inflammatory response
          immune response
          G-protein coupled receptor signaling pathway
          positive regulation of cell proliferation
          response to cold
          response to virus
          response to gamma radiation
          T cell chemotaxis
          response to auditory stimulus
          negative regulation of angiogenesis
          positive regulation of cell migration
          positive regulation of cAMP metabolic process
          response to lipopolysaccharide
          response to vitamin D
          cellular response to heat
          regulation of cell proliferation
          positive regulation of cAMP-mediated signaling
          negative regulation of myoblast differentiation
          positive regulation of release of sequestered calcium ion into cytosol
          defense response to virus
          chemokine-mediated signaling pathway
          cellular response to lipopolysaccharide
          negative regulation of myoblast fusion
          positive regulation of T cell migration
          cytokine activity
          chemokine activity
          heparin binding
          CXCR3 chemokine receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1039128 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1039128 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1039128 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1039128 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1039128 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701591071 - -
          Agilent Rat cDNA Microarray (G4105A) 701591071 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10771658 - -
          Affymetrix Rat Gene 1.0 ST Array 10771656 - -
          Affymetrix Rat Gene 1.0 ST Array 10771657 - -
          Affymetrix Rat Gene 1.0 ST Array 10771659 - -
          Affymetrix Rat Gene 1.1 ST Array 10771655 - -
          Rat Genome 230 2.0 Array 1387969_at - -
          Rat Genome 230 2.0 Array 1387969_PM_at - -
          Affymetrix HT Rat Focus Array 1387969_at - -
          Affymetrix HT Rat Focus Array 1387969_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1400001980.rn.1 TC1400000252.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400001979.rn.1 TC1400000252.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400001977.rn.1 TC1400000252.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400001978.rn.1 TC1400000252.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400001976.rn.1 TC1400000252.rn.1 RTA TC
          Rat Expression Set 230 1387969_PM_at - -
          Rat Expression Set 230 1387969_at - -
          Affymetrix Rat Gene 1.0 ST Array 10771655 - -
          Rat Genome U34 Set U17035_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21382174 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364335 - -
          RatRef-12 Expression BeadChip ILMN_59474 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline