Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Amtn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044296.1 | NP_001037761.1 | 7 - 8 | 450 | 58 | |
XM_008770057.1 | XP_008768279.1 | 6 - 7 | 2106 | 58 | ||
GenBank mRNA | AM231717.1 | - | 7 - 8 | 410 | 58 | |
DQ198381.1 | - | 7 - 8 | 450 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus Probe spans exons |