Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01414044_m1
          See other G3BP2 GE Assays ›
          Gene Symbol:
          G3bp2
          Gene Name:
          GTPase activating protein (SH3 domain) binding protein 2
          Gene Aliases:
          RGD1309571
          Chromosome Location:
          Chr.14: 17437145 - 17523254 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01414044_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: G3bp2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013989.1 NP_001014011.1 4 - 5 729 152
          XM_006250733.2 XP_006250795.1 4 - 5 723 152
          XM_006250734.2 XP_006250796.1 4 - 5 778 152
          XM_006250735.1 XP_006250797.1 4 - 5 561 152
          XM_006250736.2 XP_006250798.1 3 - 4 372 152
          GenBank mRNA BC079225.1 - 4 - 5 729 7132309 152
          FQ212261.1 - 4 - 5 566 152

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400000259.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene G3bp2

          Panther Classification:

          Molecular Function -

          RNA metabolism protein nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          nucleotide binding
          nucleic acid binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P23479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 200955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274916 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91899 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 381902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540252 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 576661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945113 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 635199 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440561 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850497 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141318 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 9632 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170781 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 140290 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P079058 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P079058 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P079058 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P23479 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77660 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73161 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10771721 - -
          Affymetrix Rat Gene 1.0 ST Array 10771726 - -
          Affymetrix Rat Gene 1.0 ST Array 10771724 - -
          Affymetrix Rat Gene 1.0 ST Array 10771728 - -
          Affymetrix Rat Gene 1.0 ST Array 10771729 - -
          Affymetrix Rat Gene 1.0 ST Array 10771727 - -
          Affymetrix Rat Gene 1.0 ST Array 10771725 - -
          Affymetrix Rat Gene 1.0 ST Array 10771723 - -
          Affymetrix Rat Gene 1.0 ST Array 10771722 - -
          Affymetrix Rat Gene 1.1 ST Array 10771720 - -
          Rat Genome 230 2.0 Array 1375974_PM_at - -
          Rat Genome 230 2.0 Array 1375974_at - -
          Rat Genome 230 2.0 Array 1374501_PM_at - -
          Rat Genome 230 2.0 Array 1374501_at - -
          Affymetrix Rat Clariom S Assay PSR1400002060.rn.1 TC1400000259.rn.1 RTA TC
          Rat Expression Set 230 1374501_at - -
          Rat Expression Set 230 1374501_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10771720 - -
          Rat Genome U34 Set rc_AA894119_at - -
          Rat Genome U34 Set rc_AA892554_g_at - -
          Rat Genome U34 Set rc_AA892554_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21043112 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1530348 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355444 - -
          RatRef-12 Expression BeadChip ILMN_160852 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline