Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01414083_m1
          See other CDKL2 GE Assays ›
          Gene Symbol:
          Cdkl2
          Gene Name:
          cyclin-dependent kinase-like 2 (CDC2-related kinase)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 17530166 - 17565950 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01414083_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cdkl2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012035.1 NP_001012035.1 8 - 9 1514 147
          XM_006250737.2 XP_006250799.1 8 - 9 1642 147
          XM_006250738.2 XP_006250800.1 9 - 10 1751 147
          XM_006250739.2 XP_006250801.1 8 - 9 2142 147
          XM_006250740.2 XP_006250802.1 9 - 10 2203 147
          XM_006250741.2 XP_006250803.1 8 - 9 2094 147
          XM_008770028.1 XP_008768250.1 9 - 10 2251 147
          XM_008770029.1 XP_008768251.1 8 - 9 2098 147
          GenBank mRNA AY383681.1 - 8 - 9 1207 147
          BC083590.1 - 8 - 9 1514 7132550 147

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400000261.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cdkl2

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          regulation of cell cycle
          protein serine/threonine kinase activity
          cyclin-dependent protein serine/threonine kinase activity
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P038747 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 482142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580968 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 678309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 367950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 209585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 803666 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 53756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935179 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961882 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 319145 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907919 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90817 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 122771 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P038747 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P038747 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P038747 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P330318 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24461 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74683 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2848 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10771735 - -
          Affymetrix Rat Gene 1.0 ST Array 10771733 - -
          Affymetrix Rat Gene 1.0 ST Array 10771737 - -
          Affymetrix Rat Gene 1.0 ST Array 10771743 - -
          Affymetrix Rat Gene 1.0 ST Array 10771734 - -
          Affymetrix Rat Gene 1.0 ST Array 10771742 - -
          Affymetrix Rat Gene 1.0 ST Array 10771744 - -
          Affymetrix Rat Gene 1.0 ST Array 10771745 - -
          Affymetrix Rat Gene 1.0 ST Array 10771739 - -
          Affymetrix Rat Gene 1.0 ST Array 10771738 - -
          Affymetrix Rat Gene 1.0 ST Array 10771736 - -
          Affymetrix Rat Gene 1.0 ST Array 10771746 - -
          Affymetrix Rat Gene 1.1 ST Array 10771732 - -
          Rat Genome 230 2.0 Array 1389736_at - -
          Rat Genome 230 2.0 Array 1389736_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1400002096.rn.1 TC1400000261.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10771732 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22203032 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376064 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376710 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline