Search
Search

Adamts3| Gene Symbol: Adamts3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107212.1 | NP_001100682.1 | 13 - 14 | 2369 | 66 | |
| XM_006250773.2 | XP_006250835.1 | 13 - 14 | 2369 | 66 | ||
| XM_006250774.2 | XP_006250836.1 | 13 - 14 | 1994 | 66 | ||
Set Membership: |
|