Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01414656_m1
          See other CDKN3 GE Assays ›
          Gene Symbol:
          Cdkn3
          Gene Name:
          cyclin-dependent kinase inhibitor 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 23553168 - 23564538 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01414656_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cdkn3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106028.1 NP_001099498.1 2 - 3 156 71
          XM_006251791.2 XP_006251853.1 2 - 3 187 71
          XM_006251792.2 XP_006251854.1 2 - 3 187 71
          GenBank mRNA BC167026.1 - 1 - 2 101 8375927 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500000230.rn.1

          Set Membership:

          Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          protein phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          cell cycle arrest
          biological_process
          peptidyl-tyrosine dephosphorylation
          molecular_function
          protein serine/threonine phosphatase activity
          protein tyrosine phosphatase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P001060 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 236370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159332 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203882 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 109735 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P001060 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P001060 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P001060 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P513360 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10779640 - -
          Affymetrix Rat Gene 1.0 ST Array 10779642 - -
          Affymetrix Rat Gene 1.0 ST Array 10779639 - -
          Affymetrix Rat Gene 1.0 ST Array 10779645 - -
          Affymetrix Rat Gene 1.0 ST Array 10779643 - -
          Affymetrix Rat Gene 1.0 ST Array 10779644 - -
          Affymetrix Rat Gene 1.0 ST Array 10779641 - -
          Affymetrix Rat Gene 1.1 ST Array 10779638 - -
          Rat Genome 230 2.0 Array 1372685_at - -
          Rat Genome 230 2.0 Array 1372685_PM_at - -
          Affymetrix HT Rat Focus Array 1372685_at - -
          Affymetrix HT Rat Focus Array 1372685_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1500001582.rn.1 TC1500000230.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500001583.rn.1 TC1500000230.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500001580.rn.1 TC1500000230.rn.1 RTA TC
          Rat Expression Set 230 1372685_at - -
          Rat Expression Set 230 1372685_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10779638 - -
          Rat Genome U34 Set rc_AI230798_at - -
          Rat Genome U34 Set rc_AI072559_f_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20737395 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364508 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline