Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01414698_m1
          See other NDRG2 GE Assays ›
          Gene Symbol:
          Ndrg2
          Gene Name:
          NDRG family member 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 28305820 - 28314459 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01414698_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ndrg2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001270862.1 NP_001257791.1 6 - 7 478 102
          NM_001270863.1 NP_001257792.1 6 - 7 460 102
          NM_001270864.1 NP_001257793.1 5 - 6 418 102
          NM_133583.2 NP_598267.2 5 - 6 436 102
          GenBank mRNA AF334105.1 - 6 - 7 442 102
          AF334106.1 - 5 - 6 400 102
          AJ426424.1 - 6 - 7 491 102
          AJ426425.1 - 5 - 6 449 102
          AJ426426.1 - 6 - 7 454 102
          AJ426427.1 - 5 - 6 412 102
          FM045410.1 - 6 - 7 478 102
          FM057759.1 - 5 - 6 418 102

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500001577.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ndrg2

          Panther Classification:

          Molecular Function -

          serine protease

          Gene Ontology Categories:

          Function(s)

          negative regulation of cytokine production
          signal transduction
          regulation of vascular endothelial growth factor production
          Wnt signaling pathway
          substantia nigra development
          cell differentiation
          negative regulation of smooth muscle cell proliferation
          negative regulation of ERK1 and ERK2 cascade
          regulation of platelet-derived growth factor production

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P034680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591463 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 40894 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10010 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P034680 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P034680 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P034680 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P547616 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78706 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24350 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10783226 - -
          Affymetrix Rat Gene 1.0 ST Array 10783215 - -
          Affymetrix Rat Gene 1.0 ST Array 10783227 - -
          Affymetrix Rat Gene 1.0 ST Array 10783221 - -
          Affymetrix Rat Gene 1.0 ST Array 10783216 - -
          Affymetrix Rat Gene 1.0 ST Array 10783224 - -
          Affymetrix Rat Gene 1.0 ST Array 10783220 - -
          Affymetrix Rat Gene 1.0 ST Array 10783223 - -
          Affymetrix Rat Gene 1.0 ST Array 10783218 - -
          Affymetrix Rat Gene 1.0 ST Array 10783229 - -
          Affymetrix Rat Gene 1.0 ST Array 10783222 - -
          Affymetrix Rat Gene 1.0 ST Array 10783219 - -
          Affymetrix Rat Gene 1.0 ST Array 10783214 - -
          Affymetrix Rat Gene 1.0 ST Array 10783217 - -
          Affymetrix Rat Gene 1.0 ST Array 10783228 - -
          Affymetrix Rat Gene 1.0 ST Array 10783225 - -
          Affymetrix Rat Gene 1.1 ST Array 10783213 - -
          Rat Genome 230 2.0 Array 1387121_a_at - -
          Rat Genome 230 2.0 Array 1398111_PM_at - -
          Rat Genome 230 2.0 Array 1387121_PM_a_at - -
          Rat Genome 230 2.0 Array 1398111_at - -
          Affymetrix HT Rat Focus Array 1387121_PM_a_at - -
          Affymetrix HT Rat Focus Array 1387121_a_at - -
          Affymetrix Rat Clariom S Assay PSR1500009984.rn.1 TC1500001577.rn.1 RTA TC
          Rat Expression Set 230 1387121_PM_a_at - -
          Rat Expression Set 230 1387121_a_at - -
          Rat Expression Set 230 1398111_PM_at - -
          Rat Expression Set 230 1398111_at - -
          Affymetrix Rat Gene 1.0 ST Array 10783213 - -
          Rat Genome U34 Set rc_AA799560_at - -
          Rat Genome U34 Set rc_AI170696_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21691637 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356551 - -
          RatRef-12 Expression BeadChip ILMN_64748 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline