Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01414796_m1
          See other METTL3 GE Assays ›
          Gene Symbol:
          Mettl3
          Gene Name:
          methyltransferase-like 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 28710299 - 28721500 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01414796_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mettl3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024794.1 NP_001019965.1 3 - 4 791 121
          XM_008770555.1 XP_008768777.1 3 - 4 1164 121
          GenBank mRNA BC097400.1 - 3 - 4 791 7453630 121

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500001587.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mettl3

          Panther Classification:

          Molecular Function -

          RNA metabolism protein RNA methyltransferase nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          RNA methylation
          mRNA processing
          circadian rhythm
          stem cell population maintenance
          mRNA destabilization
          mRNA methylation
          mRNA (2'-O-methyladenosine-N6-)-methyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P499113 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 481508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171421 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 429557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927620 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 153333 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103673 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P499113 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P499113 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P499113 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P499113 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28191 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51538 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10783333 - -
          Affymetrix Rat Gene 1.0 ST Array 10783337 - -
          Affymetrix Rat Gene 1.0 ST Array 10783332 - -
          Affymetrix Rat Gene 1.0 ST Array 10783336 - -
          Affymetrix Rat Gene 1.0 ST Array 10783339 - -
          Affymetrix Rat Gene 1.0 ST Array 10783335 - -
          Affymetrix Rat Gene 1.0 ST Array 10783342 - -
          Affymetrix Rat Gene 1.0 ST Array 10783331 - -
          Affymetrix Rat Gene 1.0 ST Array 10783338 - -
          Affymetrix Rat Gene 1.0 ST Array 10783341 - -
          Affymetrix Rat Gene 1.0 ST Array 10783340 - -
          Affymetrix Rat Gene 1.0 ST Array 10783334 - -
          Affymetrix Rat Gene 1.1 ST Array 10783330 - -
          Rat Genome 230 2.0 Array 1385149_PM_at - -
          Rat Genome 230 2.0 Array 1376986_at - -
          Rat Genome 230 2.0 Array 1376986_PM_at - -
          Rat Genome 230 2.0 Array 1385149_at - -
          Affymetrix HT Rat Focus Array 1376986_PM_at - -
          Affymetrix HT Rat Focus Array 1376986_at - -
          Affymetrix Rat Clariom S Assay PSR1500010234.rn.1 TC1500001587.rn.1 RTA TC
          Rat Expression Set 230 1376986_PM_at - -
          Rat Expression Set 230 1376986_at - -
          Affymetrix Rat Gene 1.0 ST Array 10783330 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21989655 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366172 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline