Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01415097_m1
          See other DIEXF GE Assays ›
          Gene Symbol:
          Diexf
          Gene Name:
          digestive organ expansion factor homolog (zebrafish)
          Gene Aliases:
          Def
          Chromosome Location:
          Chr.13: 111828052 - 111849654 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01415097_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Diexf
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013986.2 NP_001014008.1 11 - 12 2145 85
          XM_006250482.2 XP_006250544.1 12 - 13 2335 85
          GenBank mRNA BC087114.1 - 11 - 12 2099 7193786 85
          FQ217506.1 - 12 - 13 2318 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300002242.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Diexf

          Panther Classification:

          Molecular Function -

          RNA metabolism protein nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          maturation of SSU-rRNA from tricistronic rRNA transcript (SSU-rRNA, 5.8S rRNA, LSU-rRNA)
          multicellular organism development
          rRNA binding
          U3 snoRNA binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P047271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838795 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 429558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17547 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 322905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 235849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 965399 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505447 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 611587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801747 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 102826 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 34867 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171006 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P047271 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P047271 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P047271 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P333550 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 20100 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81681 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93060 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10770775 - -
          Affymetrix Rat Gene 1.0 ST Array 10770767 - -
          Affymetrix Rat Gene 1.0 ST Array 10770765 - -
          Affymetrix Rat Gene 1.0 ST Array 10770771 - -
          Affymetrix Rat Gene 1.0 ST Array 10770766 - -
          Affymetrix Rat Gene 1.0 ST Array 10770777 - -
          Affymetrix Rat Gene 1.0 ST Array 10770769 - -
          Affymetrix Rat Gene 1.0 ST Array 10770774 - -
          Affymetrix Rat Gene 1.0 ST Array 10770772 - -
          Affymetrix Rat Gene 1.0 ST Array 10770776 - -
          Affymetrix Rat Gene 1.0 ST Array 10770768 - -
          Affymetrix Rat Gene 1.0 ST Array 10770770 - -
          Affymetrix Rat Gene 1.0 ST Array 10770773 - -
          Affymetrix Rat Gene 1.1 ST Array 10770764 - -
          Rat Genome 230 2.0 Array 1390035_at - -
          Rat Genome 230 2.0 Array 1390035_PM_at - -
          Affymetrix HT Rat Focus Array 1390035_PM_at - -
          Affymetrix HT Rat Focus Array 1390035_at - -
          Affymetrix Rat Clariom S Assay PSR1300015968.rn.1 TC1300002242.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300015969.rn.1 TC1300002242.rn.1 RTA TC
          Rat Expression Set 230 1390035_PM_at - -
          Rat Expression Set 230 1390035_at - -
          Affymetrix Rat Gene 1.0 ST Array 10770764 - -
          Rat Genome U34 Set rc_AI008964_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359162 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline