Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01415287_m1
          See other EPRS GE Assays ›
          Gene Symbol:
          Eprs
          Gene Name:
          glutamyl-prolyl-tRNA synthetase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 103300920 - 103371651 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01415287_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Eprs
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024238.1 NP_001019409.1 31 - 32 4309 68
          XM_006250425.2 XP_006250487.1 32 - 33 4562 68
          XM_006250426.2 XP_006250488.1 31 - 32 4541 68
          XM_006250427.2 XP_006250489.1 28 - 29 3942 68
          GenBank mRNA AY383705.1 - 31 - 32 4309 68
          BC088324.1 - 11 - 12 1361 68
          FQ220944.1 - 7 - 8 863 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300000978.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Eprs

          Panther Classification:

          Molecular Function -

          aminoacyl-tRNA synthetase

          Gene Ontology Categories:

          Function(s) Process(es)

          prolyl-tRNA aminoacylation
          negative regulation of translation
          cellular response to interferon-gamma
          proline-tRNA ligase activity
          ATP binding
          RNA stem-loop binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P079650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 319572 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 641227 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556894 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 863401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 943540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 561407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 309370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827098 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137678 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 678821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 639227 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368140 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 795991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826242 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180762 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 200708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809549 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 426552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 275607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522897 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400445 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 367841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 531472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 344284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953394 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 43773 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353572 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595717 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 77032 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 34362 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 2180 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50951 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167004 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136078 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 18597 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P079650 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P079650 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P079650 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1000389 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 16908 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 69804 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78585 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94995 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103091 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10766540 - -
          Affymetrix Rat Gene 1.0 ST Array 10766546 - -
          Affymetrix Rat Gene 1.0 ST Array 10766531 - -
          Affymetrix Rat Gene 1.0 ST Array 10766545 - -
          Affymetrix Rat Gene 1.0 ST Array 10766555 - -
          Affymetrix Rat Gene 1.0 ST Array 10766538 - -
          Affymetrix Rat Gene 1.0 ST Array 10766543 - -
          Affymetrix Rat Gene 1.0 ST Array 10766550 - -
          Affymetrix Rat Gene 1.0 ST Array 10766529 - -
          Affymetrix Rat Gene 1.0 ST Array 10766548 - -
          Affymetrix Rat Gene 1.0 ST Array 10766523 - -
          Affymetrix Rat Gene 1.0 ST Array 10766535 - -
          Affymetrix Rat Gene 1.0 ST Array 10766547 - -
          Affymetrix Rat Gene 1.0 ST Array 10766552 - -
          Affymetrix Rat Gene 1.0 ST Array 10766532 - -
          Affymetrix Rat Gene 1.0 ST Array 10766553 - -
          Affymetrix Rat Gene 1.0 ST Array 10766551 - -
          Affymetrix Rat Gene 1.0 ST Array 10766539 - -
          Affymetrix Rat Gene 1.0 ST Array 10766527 - -
          Affymetrix Rat Gene 1.0 ST Array 10766549 - -
          Affymetrix Rat Gene 1.0 ST Array 10766526 - -
          Affymetrix Rat Gene 1.0 ST Array 10766542 - -
          Affymetrix Rat Gene 1.0 ST Array 10766534 - -
          Affymetrix Rat Gene 1.0 ST Array 10766544 - -
          Affymetrix Rat Gene 1.0 ST Array 10766554 - -
          Affymetrix Rat Gene 1.0 ST Array 10766541 - -
          Affymetrix Rat Gene 1.0 ST Array 10766525 - -
          Affymetrix Rat Gene 1.0 ST Array 10766522 - -
          Affymetrix Rat Gene 1.0 ST Array 10766537 - -
          Affymetrix Rat Gene 1.0 ST Array 10766528 - -
          Affymetrix Rat Gene 1.0 ST Array 10766536 - -
          Affymetrix Rat Gene 1.0 ST Array 10766524 - -
          Affymetrix Rat Gene 1.1 ST Array 10766521 - -
          Rat Genome 230 2.0 Array 1373418_at - -
          Rat Genome 230 2.0 Array 1383455_PM_at - -
          Rat Genome 230 2.0 Array 1382648_at - -
          Rat Genome 230 2.0 Array 1382040_at - -
          Rat Genome 230 2.0 Array 1382648_PM_at - -
          Rat Genome 230 2.0 Array 1373418_PM_at - -
          Rat Genome 230 2.0 Array 1383455_at - -
          Rat Genome 230 2.0 Array 1382040_PM_at - -
          Affymetrix HT Rat Focus Array 1382648_at - -
          Affymetrix HT Rat Focus Array 1382040_at - -
          Affymetrix HT Rat Focus Array 1382648_PM_at - -
          Affymetrix HT Rat Focus Array 1383455_at - -
          Affymetrix HT Rat Focus Array 1373418_PM_at - -
          Affymetrix HT Rat Focus Array 1383455_PM_at - -
          Affymetrix HT Rat Focus Array 1373418_at - -
          Affymetrix HT Rat Focus Array 1382040_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1300007085.rn.1 TC1300000978.rn.1 RTA TC
          Rat Expression Set 230 1373418_at - -
          Rat Expression Set 230 1373418_PM_at - -
          Rat Expression Set 230 1382648_at - -
          Rat Expression Set 230 1382648_PM_at - -
          Rat Expression Set 230 1383455_PM_at - -
          Rat Expression Set 230 1382040_at - -
          Rat Expression Set 230 1382040_PM_at - -
          Rat Expression Set 230 1383455_at - -
          Affymetrix Rat Gene 1.0 ST Array 10766521 - -
          Rat Genome U34 Set rc_AI069984_at - -
          Rat Genome U34 Set rc_AA964944_at - -
          Rat Genome U34 Set rc_AI029450_at - -
          Rat Genome U34 Set rc_AI235566_at - -
          Rat Genome U34 Set rc_AI234919_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20776093 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_2038894 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline