Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Kiz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006235181.2 | XP_006235243.1 | 5 - 6 | 1164 | 71 | |
XM_006235182.2 | XP_006235244.1 | 5 - 6 | 1164 | 71 | ||
XM_008762291.1 | XP_008760513.1 | 5 - 6 | 1164 | 71 | ||
XM_008762292.1 | XP_008760514.1 | 2 - 3 | 935 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Probe spans exons |