Search
Search
| Gene Symbol: Banf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006235132.2 | XP_006235194.1 | 3 - 4 | 362 | 102 | |
| XM_006235134.2 | XP_006235196.1 | 5 - 6 | 1267 | 102 | ||
| XM_006235135.2 | XP_006235197.1 | 3 - 4 | 1056 | 102 | ||
| XM_006235137.2 | XP_006235199.1 | 2 - 3 | 204 | 102 | ||
| XM_008762281.1 | XP_008760503.1 | 2 - 3 | 326 | 102 | ||
| XM_008762282.1 | XP_008760504.1 | 3 - 4 | 1012 | 102 | ||
| XM_215865.7 | XP_215865.3 | 2 - 3 | 937 | 102 | ||
Set Membership: |
|