Search
Search
| Gene Symbol: Snx5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106518.1 | NP_001099988.1 | 6 - 7 | 655 | 124 | |
| XM_006235119.2 | XP_006235181.1 | 6 - 7 | 705 | 124 | ||
| GenBank mRNA | BC160883.1 | - | 6 - 7 | 636 | 7382263 | 124 |
| FQ231966.1 | - | 6 - 7 | 637 | 124 | ||
Set Membership: |
|