Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01415916_g1
          See other RGS19 GE Assays ›
          Gene Symbol:
          Rgs19
          Gene Name:
          regulator of G-protein signaling 19
          Gene Aliases:
          Camki, Gaip
          Chromosome Location:
          Chr.3: 177196625 - 177204806 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01415916_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rgs19
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021661.2 NP_067693.1 5 - 6 755 87
          XM_006235801.2 XP_006235863.1 5 - 6 3516 87
          XM_008762540.1 XP_008760762.1 4 - 5 1028 87
          XM_008762541.1 XP_008760763.1 4 - 5 495 87
          XM_008762542.1 XP_008760764.1 4 - 5 491 87
          GenBank mRNA AF068136.1 - 4 - 5 463 87
          BC088141.1 - 5 - 6 755 7377022 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300005063.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rgs19

          Panther Classification:

          Molecular Function -

          GTPase-activating protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          regulation of G-protein coupled receptor protein signaling pathway
          termination of G-protein coupled receptor signaling pathway
          positive regulation of GTPase activity
          G-protein alpha-subunit binding
          GTPase activator activity
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P380441 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638768 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170627 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821008 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154615 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P380441 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P380441 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P380441 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P380441 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P472483 - -
          Agilent Rat cDNA Microarray (G4105A) 701898010 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10852602 - -
          Affymetrix Rat Gene 1.0 ST Array 10852606 - -
          Affymetrix Rat Gene 1.0 ST Array 10852604 - -
          Affymetrix Rat Gene 1.0 ST Array 10852608 - -
          Affymetrix Rat Gene 1.0 ST Array 10852605 - -
          Affymetrix Rat Gene 1.0 ST Array 10852601 - -
          Affymetrix Rat Gene 1.0 ST Array 10852607 - -
          Affymetrix Rat Gene 1.0 ST Array 10852603 - -
          Affymetrix Rat Gene 1.1 ST Array 10852600 - -
          Rat Genome 230 2.0 Array 1370133_PM_at - -
          Rat Genome 230 2.0 Array 1370133_at - -
          Affymetrix HT Rat Focus Array 1370133_PM_at - -
          Affymetrix HT Rat Focus Array 1370133_at - -
          Affymetrix Rat Clariom S Assay PSR0300038557.rn.1 TC0300005063.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300038560.rn.1 TC0300005063.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300038558.rn.1 TC0300005063.rn.1 RTA TC
          Rat Expression Set 230 1370133_PM_at - -
          Rat Expression Set 230 1370133_at - -
          Affymetrix Rat Gene 1.0 ST Array 10852600 - -
          Rat Genome U34 Set AF068136_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21337946 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361984 - -
          RatRef-12 Expression BeadChip ILMN_52669 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline