Search
Search
| Gene Symbol: Polr3k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014259.1 | NP_001014281.1 | 2 - 3 | 266 | 59 | |
| GenBank mRNA | BC089990.1 | - | 2 - 3 | 266 | 7319381 | 59 |
| FQ212572.1 | - | 2 - 3 | 288 | 59 | ||
| FQ213710.1 | - | 2 - 3 | 289 | 59 | ||
| FQ213830.1 | - | 2 - 3 | 300 | 59 | ||
| FQ216547.1 | - | 2 - 3 | 289 | 59 | ||
| FQ226947.1 | - | 2 - 3 | 271 | 59 | ||
| FQ232144.1 | - | 2 - 3 | 300 | 59 | ||
Set Membership: |
|