Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01416259_m1
          See other EGFL6 GE Assays ›
          Gene Symbol:
          Egfl6
          Gene Name:
          EGF-like-domain, multiple 6
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 29430791 - 29488737 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01416259_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Egfl6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108254.1 NP_001101724.1 2 - 3 389 67
          XM_008773171.1 XP_008771393.1 2 - 3 429 67
          XM_008773172.1 XP_008771394.1 2 - 3 429 67
          GenBank mRNA BC081944.1 - 2 - 3 360 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00000291.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of cell-substrate adhesion
          extracellular matrix organization
          calcium ion binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P19435 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 441721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 803701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 309593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 385489 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964218 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 702590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306045 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 901289 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752233 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1418 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294521 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 895123 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759372 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 367696 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136797 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308362 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868210 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468424 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230703 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248555 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866385 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329068 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 65175 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 80683 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 65050 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 139886 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138573 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44881 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P19435 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P19435 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P19435 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P19435 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27127 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 38263 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7383 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10933350 - -
          Affymetrix Rat Gene 1.0 ST Array 10933360 - -
          Affymetrix Rat Gene 1.0 ST Array 10933357 - -
          Affymetrix Rat Gene 1.0 ST Array 10933352 - -
          Affymetrix Rat Gene 1.0 ST Array 10933358 - -
          Affymetrix Rat Gene 1.0 ST Array 10933351 - -
          Affymetrix Rat Gene 1.0 ST Array 10933361 - -
          Affymetrix Rat Gene 1.0 ST Array 10933359 - -
          Affymetrix Rat Gene 1.0 ST Array 10933354 - -
          Affymetrix Rat Gene 1.0 ST Array 10933355 - -
          Affymetrix Rat Gene 1.0 ST Array 10933356 - -
          Affymetrix Rat Gene 1.0 ST Array 10933353 - -
          Affymetrix Rat Gene 1.1 ST Array 10933349 - -
          Rat Genome 230 2.0 Array 1393335_PM_at - -
          Rat Genome 230 2.0 Array 1393335_at - -
          Affymetrix HT Rat Focus Array 1393335_at - -
          Affymetrix HT Rat Focus Array 1393335_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0X00002176.rn.1 TC0X00000291.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00002178.rn.1 TC0X00000291.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00002183.rn.1 TC0X00000291.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00002173.rn.1 TC0X00000291.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00002190.rn.1 TC0X00000291.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00002177.rn.1 TC0X00000291.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00002172.rn.1 TC0X00000291.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00002175.rn.1 TC0X00000291.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00002189.rn.1 TC0X00000291.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00002181.rn.1 TC0X00000291.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00002187.rn.1 TC0X00000291.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00002182.rn.1 TC0X00000291.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00002186.rn.1 TC0X00000291.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00002188.rn.1 TC0X00000291.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00002179.rn.1 TC0X00000291.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00002174.rn.1 TC0X00000291.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00002180.rn.1 TC0X00000291.rn.1 RTA TC
          Rat Expression Set 230 1393335_PM_at - -
          Rat Expression Set 230 1393335_at - -
          Affymetrix Rat Gene 1.0 ST Array 10933349 - -
          Rat Genome U34 Set rc_AI070850_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361440 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline