Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01416277_m1
          See other CAR5B GE Assays ›
          Gene Symbol:
          Car5b
          Gene Name:
          carbonic anhydrase 5b, mitochondrial
          Gene Aliases:
          Ca5b
          Chromosome Location:
          Chr.X: 32232108 - 32291124 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01416277_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Car5b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001005551.2 NP_001005551.1 6 - 7 833 67
          XM_006256859.2 XP_006256921.1 7 - 8 930 67
          GenBank mRNA BC081872.1 - 6 - 7 835 7106842 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00000314.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Car5b

          Gene Ontology Categories:

          Function(s) Process(es)

          one-carbon metabolic process
          carbonate dehydratase activity
          zinc ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1005051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 379987 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 845223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154958 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 80644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91045 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 713885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 364799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216523 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 837150 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 846552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527288 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419183 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 547679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740654 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850541 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145579 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137762 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14668 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 175806 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52394 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 144660 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1005051 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1005051 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1005051 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1005051 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55576 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99070 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97261 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53614 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10933456 - -
          Affymetrix Rat Gene 1.0 ST Array 10933454 - -
          Affymetrix Rat Gene 1.0 ST Array 10933453 - -
          Affymetrix Rat Gene 1.0 ST Array 10933455 - -
          Affymetrix Rat Gene 1.0 ST Array 10933451 - -
          Affymetrix Rat Gene 1.0 ST Array 10933450 - -
          Affymetrix Rat Gene 1.0 ST Array 10933452 - -
          Affymetrix Rat Gene 1.1 ST Array 10933449 - -
          Rat Genome 230 2.0 Array 1390807_PM_at - -
          Rat Genome 230 2.0 Array 1377573_PM_at - -
          Rat Genome 230 2.0 Array 1390807_at - -
          Rat Genome 230 2.0 Array 1377573_at - -
          Affymetrix HT Rat Focus Array 1390807_PM_at - -
          Affymetrix HT Rat Focus Array 1377573_at - -
          Affymetrix HT Rat Focus Array 1390807_at - -
          Affymetrix HT Rat Focus Array 1377573_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0X00002336.rn.1 TC0X00000314.rn.1 RTA TC
          Rat Expression Set 230 1390807_PM_at - -
          Rat Expression Set 230 1390807_at - -
          Rat Expression Set 230 1377573_at - -
          Rat Expression Set 230 1377573_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10933449 - -
          Rat Genome U34 Set rc_AA892953_s_at - -
          Rat Genome U34 Set rc_AA848689_at - -
          Rat Genome U34 Set rc_AA925081_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22308508 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356489 - -
          RatRef-12 Expression BeadChip ILMN_50870 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline