Search
Search

Shroom2| Gene Symbol: Shroom2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047893.3 | NP_001041358.2 | 9 - 10 | 4208 | 130 | |
| XM_006256816.2 | XP_006256878.1 | 9 - 10 | 4290 | 130 | ||
| XM_006256817.2 | XP_006256879.1 | 9 - 10 | 4287 | 130 | ||
| XM_008773126.1 | XP_008771348.1 | 8 - 9 | 4173 | 130 | ||
| XM_008773127.1 | XP_008771349.1 | 5 - 6 | 1765 | 130 | ||
| XM_008773128.1 | XP_008771350.1 | 5 - 6 | 3433 | 130 | ||
| XM_008773129.1 | XP_008771351.1 | 5 - 6 | 3415 | 130 | ||
| XM_008773130.1 | XP_008771352.1 | 5 - 6 | 3418 | 130 | ||
| XM_008773131.1 | XP_008771353.1 | 5 - 6 | 1747 | 130 | ||
| XM_008773132.1 | XP_008771354.1 | 5 - 6 | 1750 | 130 | ||
| GenBank mRNA | AY325213.1 | - | 7 - 8 | 4004 | 130 | |
| FQ211812.1 | - | 5 - 6 | 1314 | 130 | ||
Set Membership: |
|