Search
Search

LOC503435| Gene Symbol: LOC503435 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109366.1 | NP_001102836.1 | 1 | 541 | 82 | |
| XM_006256820.2 | XP_006256882.1 | 2 | 1232 | 82 | ||
| XM_008773133.1 | XP_008771355.1 | 2 | 778 | 82 | ||
Set Membership: |
|