Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01416441_m1
          See other EPS15 GE Assays ›
          Gene Symbol:
          Eps15
          Gene Name:
          epidermal growth factor receptor pathway substrate 15
          Gene Aliases:
          RGD1307641
          Chromosome Location:
          Chr.5: 128923809 - 129022268 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01416441_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Eps15
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001009424.1 NP_001009424.1 11 - 12 83
          GenBank mRNA AB262963.1 - 11 - 12 83
          AY190609.1 - 11 - 12 83
          FQ223380.1 - 1081 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500001187.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Eps15

          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of receptor recycling
          Golgi to endosome transport
          endocytic recycling
          regulation of cell proliferation
          clathrin coat assembly
          calcium ion binding
          protein binding
          polyubiquitin binding
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P071466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57506 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807794 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808390 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 665969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564683 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864641 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519332 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459633 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205603 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 395105 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 646039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558147 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543060 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131678 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 562230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 166078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 451033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 872791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443415 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 166039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375394 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357105 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174113 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82023 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 451389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83421 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 857246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 702380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 723368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 535224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 53804 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 755102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587878 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554444 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 121673 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 21388 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 72588 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 147805 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 37908 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 168035 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101591 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91030 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138036 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 38128 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 73828 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 155509 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 109019 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P071466 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P071466 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P071466 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P19497 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P445792 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15107 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63830 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6106 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73817 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1504 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27304 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46705 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10870919 - -
          Affymetrix Rat Gene 1.0 ST Array 10870928 - -
          Affymetrix Rat Gene 1.0 ST Array 10870911 - -
          Affymetrix Rat Gene 1.0 ST Array 10870906 - -
          Affymetrix Rat Gene 1.0 ST Array 10870926 - -
          Affymetrix Rat Gene 1.0 ST Array 10870927 - -
          Affymetrix Rat Gene 1.0 ST Array 10870922 - -
          Affymetrix Rat Gene 1.0 ST Array 10870921 - -
          Affymetrix Rat Gene 1.0 ST Array 10870907 - -
          Affymetrix Rat Gene 1.0 ST Array 10870916 - -
          Affymetrix Rat Gene 1.0 ST Array 10870908 - -
          Affymetrix Rat Gene 1.0 ST Array 10870905 - -
          Affymetrix Rat Gene 1.0 ST Array 10870910 - -
          Affymetrix Rat Gene 1.0 ST Array 10870923 - -
          Affymetrix Rat Gene 1.0 ST Array 10870914 - -
          Affymetrix Rat Gene 1.0 ST Array 10870915 - -
          Affymetrix Rat Gene 1.0 ST Array 10870918 - -
          Affymetrix Rat Gene 1.0 ST Array 10870909 - -
          Affymetrix Rat Gene 1.0 ST Array 10870912 - -
          Affymetrix Rat Gene 1.0 ST Array 10870903 - -
          Affymetrix Rat Gene 1.0 ST Array 10870917 - -
          Affymetrix Rat Gene 1.0 ST Array 10870904 - -
          Affymetrix Rat Gene 1.1 ST Array 10870902 - -
          Rat Genome 230 2.0 Array 1395548_PM_at - -
          Rat Genome 230 2.0 Array 1399152_at - -
          Rat Genome 230 2.0 Array 1395548_at - -
          Rat Genome 230 2.0 Array 1399152_PM_at - -
          Affymetrix HT Rat Focus Array 1399152_at - -
          Affymetrix HT Rat Focus Array 1399152_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500008053.rn.1 TC0500001187.rn.1 RTA TC
          Rat Expression Set 230 1399152_PM_at - -
          Rat Expression Set 230 1399152_at - -
          Affymetrix Rat Gene 1.0 ST Array 10870902 - -
          Rat Genome U34 Set rc_AI059428_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21783009 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358902 - -
          RatRef-12 Expression BeadChip ILMN_68936 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline