Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01416579_m1
          See other YIPF1 GE Assays ›
          Gene Symbol:
          Yipf1
          Gene Name:
          Yip1 domain family, member 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 126932738 - 126968110 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01416579_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Yipf1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_199383.2 NP_955415.1 3 - 4 314 75
          XM_006238504.2 XP_006238566.1 3 - 4 352 75
          XM_006238505.2 XP_006238567.1 3 - 4 351 75
          GenBank mRNA BC062239.1 - 3 - 4 318 6887537 75
          FQ218908.1 - 3 - 4 313 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500001142.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Yipf1

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P081183 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749603 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727110 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 561540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949501 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220771 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 641781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132426 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 420177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220630 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362227 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 309756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 24445 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770058 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 376886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110949 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375001 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100049 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57745 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 25138 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 78295 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 24445 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 78094 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163155 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 78261 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P081183 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P081183 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P081183 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P118649 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1485 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 60541 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35612 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10870673 - -
          Affymetrix Rat Gene 1.0 ST Array 10870674 - -
          Affymetrix Rat Gene 1.0 ST Array 10870670 - -
          Affymetrix Rat Gene 1.0 ST Array 10870677 - -
          Affymetrix Rat Gene 1.0 ST Array 10870675 - -
          Affymetrix Rat Gene 1.0 ST Array 10870669 - -
          Affymetrix Rat Gene 1.0 ST Array 10870676 - -
          Affymetrix Rat Gene 1.0 ST Array 10870668 - -
          Affymetrix Rat Gene 1.0 ST Array 10870672 - -
          Affymetrix Rat Gene 1.0 ST Array 10870671 - -
          Affymetrix Rat Gene 1.1 ST Array 10870667 - -
          Rat Genome 230 2.0 Array 1372893_at - -
          Rat Genome 230 2.0 Array 1372893_PM_at - -
          Affymetrix HT Rat Focus Array 1372893_PM_at - -
          Affymetrix HT Rat Focus Array 1372893_at - -
          Affymetrix Rat Clariom S Assay PSR0500007593.rn.1 TC0500001142.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500007594.rn.1 TC0500001142.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500007583.rn.1 TC0500001142.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500007578.rn.1 TC0500001142.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500007581.rn.1 TC0500001142.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500007586.rn.1 TC0500001142.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500007597.rn.1 TC0500001142.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500007589.rn.1 TC0500001142.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500007588.rn.1 TC0500001142.rn.1 RTA TC
          Rat Expression Set 230 1372893_PM_at - -
          Rat Expression Set 230 1372893_at - -
          Affymetrix Rat Gene 1.0 ST Array 10870667 - -
          Rat Genome U34 Set rc_AI045003_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21986774 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374597 - -
          RatRef-12 Expression BeadChip ILMN_66933 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline