Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01416741_m1
          See other LRRC42 GE Assays ›
          Gene Symbol:
          Lrrc42
          Gene Name:
          leucine rich repeat containing 42
          Gene Aliases:
          RGD1308919
          Chromosome Location:
          Chr.5: 126835170 - 126856283 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01416741_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lrrc42
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025653.1 NP_001020824.1 7 - 8 1202 72
          XM_006238503.2 XP_006238565.1 7 - 8 1124 72
          GenBank mRNA BC098685.1 - 7 - 8 1202 7385408 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500003303.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lrrc42

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P041902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 562003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559299 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 10571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341771 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779565 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 646300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 429939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772050 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336399 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137699 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 36907 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 20572 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P041902 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P041902 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P041902 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P609263 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6235 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 52910 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95788 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103132 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10878547 - -
          Affymetrix Rat Gene 1.0 ST Array 10878544 - -
          Affymetrix Rat Gene 1.0 ST Array 10878541 - -
          Affymetrix Rat Gene 1.0 ST Array 10878542 - -
          Affymetrix Rat Gene 1.0 ST Array 10878540 - -
          Affymetrix Rat Gene 1.0 ST Array 10878545 - -
          Affymetrix Rat Gene 1.0 ST Array 10878543 - -
          Affymetrix Rat Gene 1.0 ST Array 10878546 - -
          Affymetrix Rat Gene 1.1 ST Array 10878539 - -
          Rat Genome 230 2.0 Array 1371879_PM_at - -
          Rat Genome 230 2.0 Array 1371879_at - -
          Affymetrix HT Rat Focus Array 1371879_PM_at - -
          Affymetrix HT Rat Focus Array 1371879_at - -
          Affymetrix Rat Clariom S Assay PSR0500023989.rn.1 TC0500003303.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500023991.rn.1 TC0500003303.rn.1 RTA TC
          Rat Expression Set 230 1371879_PM_at - -
          Rat Expression Set 230 1371879_at - -
          Affymetrix Rat Gene 1.0 ST Array 10878539 - -
          Rat Genome U34 Set rc_AA925289_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22040920 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353898 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline