Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01416883_m1
          See other ELAVL4 GE Assays ›
          Gene Symbol:
          Elavl4
          Gene Name:
          ELAV like neuron-specific RNA binding protein 4
          Gene Aliases:
          HuD, RGD1561943, r-HuD
          Chromosome Location:
          Chr.5: 130001387 - 130144513 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01416883_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Elavl4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001077651.1 NP_001071119.1 5 - 6 840 156
          XM_006238591.2 XP_006238653.1 5 - 6 1001 156
          XM_006238592.2 XP_006238654.1 5 - 6 1316 156
          XM_006238593.2 XP_006238655.1 5 - 6 749 156
          XM_006238594.2 XP_006238656.1 5 - 6 1025 156
          XM_006238595.2 XP_006238657.1 5 - 6 1000 156
          XM_008763893.1 XP_008762115.1 5 - 6 927 156
          GenBank mRNA BC158558.1 - 5 - 6 851 8371379 156
          S83320.1 - 5 - 6 840 156

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500003363.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons

          Panther Classification:

          Molecular Function -

          translation initiation factor

          Gene Ontology Categories:

          Function(s) Process(es)

          nervous system development
          aging
          learning
          locomotory behavior
          associative learning
          neuron differentiation
          regeneration
          response to endoplasmic reticulum stress
          response to cocaine
          regulation of mRNA stability
          dendrite morphogenesis
          positive regulation of dendrite development
          cellular response to nerve growth factor stimulus
          nucleotide binding
          mRNA binding
          mRNA 3'-UTR binding
          protein binding
          AU-rich element binding

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P011854 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945544 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 777345 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 617718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686977 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136187 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 619266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325220 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 299913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 757894 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218772 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546267 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221521 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 729560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195897 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955565 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 675525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197506 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 803817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 678643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 222617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 754071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 206106 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156999 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518038 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 784015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6358 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406596 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 98735 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 22164 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111134 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 80396 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 96643 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164420 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 16854 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 21430 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P011854 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P011854 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P011854 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P941612 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P792864 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P485453 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P116489 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701188534 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61110 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57391 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 50815 - -
          Agilent Rat cDNA Microarray (G4105A) 701188534 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10878720 - -
          Affymetrix Rat Gene 1.0 ST Array 10878715 - -
          Affymetrix Rat Gene 1.0 ST Array 10878719 - -
          Affymetrix Rat Gene 1.0 ST Array 10878714 - -
          Affymetrix Rat Gene 1.0 ST Array 10878716 - -
          Affymetrix Rat Gene 1.0 ST Array 10878721 - -
          Affymetrix Rat Gene 1.0 ST Array 10878718 - -
          Affymetrix Rat Gene 1.0 ST Array 10878713 - -
          Affymetrix Rat Gene 1.0 ST Array 10878717 - -
          Affymetrix Rat Gene 1.1 ST Array 10878712 - -
          Rat Genome 230 2.0 Array 1389975_PM_at - -
          Rat Genome 230 2.0 Array 1389975_at - -
          Affymetrix HT Rat Focus Array 1389975_PM_at - -
          Affymetrix HT Rat Focus Array 1389975_at - -
          Affymetrix Rat Clariom S Assay PSR0500024463.rn.1 TC0500003363.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500024464.rn.1 TC0500003363.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500024465.rn.1 TC0500003363.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500024466.rn.1 TC0500003363.rn.1 RTA TC
          Rat Expression Set 230 1389975_at - -
          Rat Expression Set 230 1389975_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10878712 - -
          Rat Genome U34 Set S83320_at - -
          Rat Genome U34 Set S83320_g_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline