Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01417562_m1
          See other THEGL GE Assays ›
          Gene Symbol:
          Thegl
          Gene Name:
          theg spermatid protein-like
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 33389511 - 33431883 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01417562_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Thegl
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001017498.1 NP_001017498.1 7 - 8 1216 136
          XM_006250846.2 XP_006250908.1 6 - 7 2005 136
          GenBank mRNA BC087065.1 - 7 - 8 1216 7132384 136

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400001628.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Thegl

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P098739 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 891521 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806795 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 713706 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787293 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341687 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 757688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390403 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 491657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829633 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628704 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 332198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267319 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 73447 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297704 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240945 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149944 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108786 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 28734 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 102240 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6025 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P098739 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P098739 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P098739 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P816426 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62098 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8063 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4847 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10776330 - -
          Affymetrix Rat Gene 1.0 ST Array 10776328 - -
          Affymetrix Rat Gene 1.0 ST Array 10776332 - -
          Affymetrix Rat Gene 1.0 ST Array 10776327 - -
          Affymetrix Rat Gene 1.0 ST Array 10776333 - -
          Affymetrix Rat Gene 1.0 ST Array 10776326 - -
          Affymetrix Rat Gene 1.0 ST Array 10776331 - -
          Affymetrix Rat Gene 1.0 ST Array 10776334 - -
          Affymetrix Rat Gene 1.0 ST Array 10776329 - -
          Affymetrix Rat Gene 1.1 ST Array 10776325 - -
          Affymetrix Rat Clariom S Assay PSR1400011906.rn.1 TC1400001628.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10776325 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21546282 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356210 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline