Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01417686_m1
          See other SIN3A GE Assays ›
          Gene Symbol:
          Sin3a
          Gene Name:
          SIN3 transcription regulator family member A
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 61748589 - 61803314 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01417686_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sin3a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108761.1 NP_001102231.1 15 - 16 2685 73
          XM_006243129.2 XP_006243191.1 15 - 16 2681 73
          XM_006243131.2 XP_006243193.1 14 - 15 2468 73
          XM_008766298.1 XP_008764520.1 14 - 15 2873 73
          XM_008766299.1 XP_008764521.1 15 - 16 2956 73
          XM_008766300.1 XP_008764522.1 15 - 16 2465 73
          XM_008766301.1 XP_008764523.1 14 - 15 2399 73
          XM_008766302.1 XP_008764524.1 14 - 15 3267 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800000781.rn.1

          Set Membership:

          Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          chromatin/chromatin-binding, or -regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          in utero embryonic development
          activation of innate immune response
          positive regulation of defense response to virus by host
          hematopoietic progenitor cell differentiation
          DNA replication
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          protein deacetylation
          aging
          response to organonitrogen compound
          regulation of hormone levels
          positive regulation of G2/M transition of mitotic cell cycle
          histone deacetylation
          positive regulation of chromatin silencing
          cellular protein localization
          negative regulation of circadian rhythm
          negative regulation of apoptotic process
          regulation of transcription from RNA polymerase II promoter in response to oxidative stress
          negative regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          response to methylglyoxal
          cellular response to glucose stimulus
          negative regulation of protein localization to nucleus
          negative regulation of histone H3-K27 acetylation
          cellular response to dopamine
          negative regulation of transcription regulatory region DNA binding
          transcription regulatory region sequence-specific DNA binding
          RNA polymerase II activating transcription factor binding
          RNA polymerase II repressing transcription factor binding
          RNA polymerase II transcription corepressor activity
          DNA binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          transcription corepressor activity
          RNA binding
          histone deacetylase activity
          protein binding
          transcription factor binding
          protein complex binding
          protein deacetylase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P387246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356633 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 414435 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174882 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 551627 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781367 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 666016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 547902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87000 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496280 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717819 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47630 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7676 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 79802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892866 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383095 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137226 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566000 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 494697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 276708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 482539 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 395201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371287 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292836 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 122117 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138577 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15467 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141734 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41987 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P387246 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P387246 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P387246 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P387246 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65335 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 30656 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45976 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10910290 - -
          Affymetrix Rat Gene 1.0 ST Array 10910272 - -
          Affymetrix Rat Gene 1.0 ST Array 10910288 - -
          Affymetrix Rat Gene 1.0 ST Array 10910277 - -
          Affymetrix Rat Gene 1.0 ST Array 10910294 - -
          Affymetrix Rat Gene 1.0 ST Array 10910295 - -
          Affymetrix Rat Gene 1.0 ST Array 10910283 - -
          Affymetrix Rat Gene 1.0 ST Array 10910286 - -
          Affymetrix Rat Gene 1.0 ST Array 10910292 - -
          Affymetrix Rat Gene 1.0 ST Array 10910284 - -
          Affymetrix Rat Gene 1.0 ST Array 10910280 - -
          Affymetrix Rat Gene 1.0 ST Array 10910276 - -
          Affymetrix Rat Gene 1.0 ST Array 10910274 - -
          Affymetrix Rat Gene 1.0 ST Array 10910291 - -
          Affymetrix Rat Gene 1.0 ST Array 10910273 - -
          Affymetrix Rat Gene 1.0 ST Array 10910281 - -
          Affymetrix Rat Gene 1.0 ST Array 10910278 - -
          Affymetrix Rat Gene 1.0 ST Array 10910271 - -
          Affymetrix Rat Gene 1.0 ST Array 10910279 - -
          Affymetrix Rat Gene 1.0 ST Array 10910287 - -
          Affymetrix Rat Gene 1.0 ST Array 10910282 - -
          Affymetrix Rat Gene 1.0 ST Array 10910289 - -
          Affymetrix Rat Gene 1.0 ST Array 10910275 - -
          Affymetrix Rat Gene 1.0 ST Array 10910285 - -
          Affymetrix Rat Gene 1.1 ST Array 10910270 - -
          Rat Genome 230 2.0 Array 1392269_at - -
          Rat Genome 230 2.0 Array 1397105_PM_at - -
          Rat Genome 230 2.0 Array 1392269_PM_at - -
          Rat Genome 230 2.0 Array 1397105_at - -
          Rat Genome 230 2.0 Array 1391498_PM_at - -
          Rat Genome 230 2.0 Array 1391498_at - -
          Affymetrix HT Rat Focus Array 1392269_PM_at - -
          Affymetrix HT Rat Focus Array 1392269_at - -
          Affymetrix HT Rat Focus Array 1397105_at - -
          Affymetrix HT Rat Focus Array 1397105_PM_at - -
          Affymetrix HT Rat Focus Array 1391498_PM_at - -
          Affymetrix HT Rat Focus Array 1391498_at - -
          Affymetrix Rat Clariom S Assay PSR0800005303.rn.1 TC0800000781.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800005305.rn.1 TC0800000781.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800005304.rn.1 TC0800000781.rn.1 RTA TC
          Rat Expression Set 230 1391498_at - -
          Rat Expression Set 230 1397105_PM_at - -
          Rat Expression Set 230 1392269_at - -
          Rat Expression Set 230 1392269_PM_at - -
          Rat Expression Set 230 1391498_PM_at - -
          Rat Expression Set 230 1397105_at - -
          Affymetrix Rat Gene 1.0 ST Array 10910270 - -
          Rat Genome U34 Set rc_AI071983_at - -
          Rat Genome U34 Set rc_AI072877_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20870948 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362562 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline