Search
Search
| Gene Symbol: Dis3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008380.1 | NP_001008381.1 | 2 - 3 | 297 | 61 | |
| XM_006243261.2 | XP_006243323.1 | 2 - 3 | 400 | 61 | ||
| XM_006243262.2 | XP_006243324.1 | 2 - 3 | 204 | 61 | ||
| XM_006243263.2 | XP_006243325.1 | 2 - 3 | 729 | 61 | ||
| XM_008766311.1 | XP_008764533.1 | 2 - 3 | 400 | 61 | ||
| GenBank mRNA | BC085932.1 | - | 2 - 3 | 297 | 7112012 | 61 |
Set Membership: |
|